Petroleum spirit, 35…60 °C ACS

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

67482-540 67482-690
CA71007-928CS 902.94 CAD
CA71007-928 CA71007-932
Petroleum spirit, 35…60 °C ACS
Petroleum spirit
Boiling Pt: 35…60 °C (1013 hPa)
Storage Temperature: Ambient
MDL Number: MFCD00081849
CAS Number: 64742-49-0
EINECS: 265-151-9
UN: 1268
ADR: 3,II

Order Now

Specification Test Results

Colour (APHA) 10 max.
Boiling range 35 - 60°C
Residue after evaporation 0.001% max.
Acidity Passes Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR