Nitric acid 67 - 70%, Environmental Grade

Supplier: Anachemia
Danger

62764-320 62764-460
CA71007-854CS 773.92 CAD
CA71007-854 CA11021-138
Nitric acid 67 - 70%, Environmental Grade
Nitric acid
Formula: HNO₃
Boiling Pt: 120.5 °C (1013 hPa)
Melting Pt: –42 °C
Density: 1.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011349
CAS Number: 7697-37-2
EINECS: 231-714-2
UN: 2031
ADR: 8,II
Merck Index: 13,06608

Order Now

Specification Test Results

Assay 67 - 70%
Colour (APHA) 10 max.
Aluminum (Al) (10) 1 ppb max.
Antimony (Sb) 0.5 ppb max.
Arsenic (As) 0.5 ppb max.
Barium(Ba) 0.1 ppb max.
Beryllium (Be) 0.1 ppb max.
Bismuth (Bi 0.1 ppb max.
Boron (B) (5) 1 ppb max.
Cadmium (Cd) 0.5 ppb max.
Calcium (Ca) (50) 1 ppb max.
Cerium (Ce) 0.1 ppb max.
Cesium (Cs) 0.1 ppb max.
Chromium (Cr) 1 ppb max.
Cobalt (Co) 0.5 ppb max.
Copper (Cu) 0.5 ppb max.
Dysprosium (Dy) 0.1 ppb max.
Erbium (Er) 0.1 ppb max.
Europium (Eu) 0.1 ppb max.
Gadolinium (Gd) 0.1 ppb max.
Gallium(Ga) 0.1 ppb max.
Germanium(Ge) 0.1 ppb max.
Gold(Au) 0.1 ppb max.
Hafnium (Hf) 0.1 ppb max.
Holmium (Ho) 0.1 ppb max.
Indium (In) 0.1 ppb max.
Iron (Fe) (5) 1 ppb max.
Lanthanum (La) 0.1 ppb max.
Lead (Pb) 0.1 ppb max.
Lithium (Li) 0.1 ppb max.
Lutetium (Lu) 0.1 ppb max.
Magnesium (Mg) (5) 1 ppb max.
Manganese (Mn) 0.1 ppb max.
Mercury (Hg) 0.1 ppb max.
Molybdenum (Mo) 0.1 ppb max.
Neodymium (Nd) 0.1 ppb max.
Nickel (Ni) 0.5 ppb max.
Niobium (Nb) 0.1 ppb max.
Palladium (Pd) 0.5 ppb max.
Platinum (Pt) 0.5 ppb max.
Potassium (K) (5) 1 ppb max.
Praseodymium (Pr) 0.1 ppb max.
Rhenium (Re) 0.1 ppb max.
Rhodium(Rh) 0.5 ppb max.
Rubidium (Rb) 0.1 ppb max.
Ruthenium (Ru) 0.5 ppb max.
Samarium (Sm) 0.1 ppb max.
Scandium (Sc) 0.1 ppb max.
Selenium (Se) 1 ppb max.
Silver (Ag) 0.1 ppb max.
Sodium (Na) (50) 1 ppb max.
Strontium (Sr) 0.1 ppb max.
Tantalum (Ta) info only
Tellurium (Te) 0.1 ppb max.
Terbium (Tb) 0.1 ppb max.
Thallium (Tl) 0.1 ppb max.
Thorium (Th) 0.1 ppb max.
Thulium (Tm) 0.1 ppb max.
Tin (Sn) 0.5 ppb max.
Titanium (Ti) 0.5 ppb max.
Tungsten (W) 0.1 ppb max.
Uranium (U) 0.1 ppb max.
Vanadium (V) 0.5 ppb max.
Ytterbium (Yb) 0.1 ppb max.
Yttrium (Y) 0.1 ppb max.
Zinc (Zn) (2) 0.5 ppb max.
Zirconium (Zr) 0.1 ppb max.
Maximums stated applicable at time of bottling. Values in brackets are likely maximum values after storage in glass.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR