Nitric acid 67 - 70%, Environmental Grade Plus

Supplier: Anachemia

Synonyms: Nitric acid solution

62759-320 62759-400
CA71007-848EA 1698.67 CAD
CA71007-848 CA71007-852
Nitric acid 67 - 70%, Environmental Grade Plus
Nitric acid
Formula: HNO₃
MW: 63.01 g/mol
Boiling Pt: 120.5 °C (1013 hPa)
Melting Pt: –42 °C
Density: 1.4 g/cm³ (20 °C)
MDL Number: MFCD00011349
CAS Number: 7697-37-2
UN: 2031
ADR: 8,II
Merck Index: 13,06608

Order Now

Specification Test Results

Assay 67 - 70%
Aluminum (Al) 20 ppt max.
Antimony (Sb) 10 ppt max.
Arsenic (As) 20 ppt max.
Barium(Ba) 10 ppt max.
Beryllium (Be) 10 ppt max.
Bismuth (Bi 10 ppt max.
Boron (B) 10 ppt max.
Cadmium (Cd) 10 ppt max.
Calcium (Ca 10 ppt max.
Cerium (Ce) 10 ppt max.
Cesium (Cs) 10 ppt max.
Chromium (Cr) 10 ppt max.
Cobalt (Co) 10 ppt max.
Copper (Cu) 10 ppt max.
Dysprosium (Dy) 1 ppt max.
Erbium (Er) 1 ppt max.
Europium (Eu) 1 ppt max.
Gadolinium (Gd) 1 ppt max.
Gallium(Ga) 10 ppt max.
Germanium(Ge) 10 ppt max.
Gold(Au) 20 ppt max.
Hafnium (Hf) 10 ppt max.
Holmium (Ho) 1 ppt max.
Indium (In) 1 ppt max.
Iron (Fe) 10 ppt max.
Lanthanum (La) 1 ppt max.
Lead (Pb) 10 ppt max.
Lithium (Li) 10 ppt max.
Lutetium (Lu) 1 ppt max.
Magnesium (Mg) 10 ppt max.
Manganese (Mn) 10 ppt max.
Mercury (Hg) 50 ppt max.
Molybdenum (Mo) 10 ppt max.
Neodymium (Nd) 1 ppt max.
Nickel (Ni) 20 ppt max.
Niobium (Nb) 1 ppt max.
Palladium (Pd) 20 ppt max.
Platinum (Pt) 20 ppt max.
Potassium (K) 10 ppt max.
Praseodymium (Pr) 1 ppt max.
Rhenium (Re) 10 ppt max.
Rhodium(Rh) 10 ppt max.
Rubidium (Rb) 10 ppt max.
Ruthenium (Ru) 20 ppt max.
Samarium (Sm) 1 ppt max.
Scandium (Sc) 10 ppt max.
Selenium (Se) info only
Silver (Ag) 10 ppt max.
Sodium (Na) 10 ppt max.
Strontium (Sr) 10 ppt max.
Tantalum (Ta) info only
Tellurium (Te) 1 ppt max.
Terbium (Tb) 1 ppt max.
Thallium (Tl) 10 ppt max.
Thorium (Th) 1 ppt max.
Thulium (Tm) 1 ppt max.
Tin (Sn) 20 ppt max.
Titanium (Ti) 10 ppt max.
Tungsten (W) 10 ppt max.
Uranium (U) 1 ppt max.
Vanadium (V) 10 ppt max.
Ytterbium (Yb) 1 ppt max.
Yttrium (Y) 1 ppt max.
Zinc (Zn) 10 ppt max.
Zirconium (Zr) 10 ppt max.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR