Methyl ethyl ketone ≥99.0% ACS
|
Danger
|
Synonyms:
EK 500 Additive, Ethyl methyl ketone, 2-Butanone, Butanone, MEK
58934-360
58934-700
CA71007-776EA
276.18
CAD
CA71007-776
CA71007-780
CA71007-784
CA71007-788
Methyl ethyl ketone ≥99.0% ACS
Methyl ethyl ketone
Formula:
C₂H₅COCH₃ Boiling Pt: 79.6 °C (1013 hPa) Melting Pt: –86 °C Density: 0.8054 g/cm³ (20 °C) Flash Pt: –4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011648 CAS Number: 78-93-3 EINECS: 201-159-0 UN: 1193 ADR: 3,II REACH: 01-2119457290-43 Merck Index: 13,06097 |
|
Specification Test Results
Assay (CH3COCH2CH3) | 99.0% min. |
Colour (APHA) | 15 max. |
Residue after evaporation | 0.0025% max. |
Titrable acid | 0.0005 meq/g max. |
Water (H2O) | 0.2% max. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...