Methanol ≥99.8% ACS
Supplier: VWR
|
Danger
|
56902-532
56902-540
56902-543
56902-700
56902-940
CA71007-734CS
4459.76
CAD
CA71007-734
CA71007-736
CA71007-738
CA71007-742
CA71007-744
CA71007-748
Methanol ≥99.8% ACS
Methanol
Formula:
CH₃OH Boiling Pt: 64.6 °C (1013 hPa) Melting Pt: –98 °C Density: 0.7918 g/cm³ (20 °C) Flash Pt: 11 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00004595 CAS Number: 67-56-1 EINECS: 200-659-6 UN: 1230 ADR: 3,II REACH: 01-2119433307-44 Merck Index: 13,05984 |
|
Specification Test Results
Assay (CH3OH) | 99.8% min. |
Substances darkened by H2SO4 | Passes Test |
Substances reducing KMnO4 | Passes Test |
Colour (APHA) | 10 max. |
Water (H2O) | 0.1% max. |
Residue after evaporation | 0.001% max. |
Solubility in water | Passes Test |
Titrable acid | 0.0003 meq/g max. |
Titrable base | 0.0002 meq/g max. |
Carbonyl compounds: | |
Acetone | 0.001% max. |
Formaldehyde (as HCHO) | 0.001% max. |
Acetaldehyde | 0.001% max. |
Appearance | Clear |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...