Iodine 0.1 N in aqueous solution volumetric solution, traceable to NIST Standard Reference Material (SRM)

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

218V-03
80721-410CS 1006.03 CAD
80721-410
Iodine 0.1 N in aqueous solution volumetric solution, traceable to NIST Standard Reference Material (SRM)
Iodine
Formula: I₂
MW: 253.81 g/mol
MDL Number: MFCD00011355
CAS Number: 7553-56-2
UN: 3264
ADR: 8,II

Order Now

Specification Test Results

Normality 0.0995 - 0.1005 N
Appearance: Transparent, reddish-brown solution, no residue, precipitate, floater.
Traceable to NIST standards

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR