Hydrofluoric acid ≥70% (w/w), Technical Grade
Supplier: Anachemia
264000
2640N1
CA11020-946CS
3829.22
CAD
CA11020-946
CA11020-944
Hydrofluoric acid ≥70% (w/w), Technical Grade
Hydrofluoric acid
Formula:
HF MW: 20.01 g/mol |
MDL Number:
MFCD00011346 CAS Number: 7664-39-3 UN: 1790 ADR: 8,I |
Specification Test Results
Assay (HF) | 70.0% w/w min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...