Hydrofluoric acid 47 - 51%, Environmental Grade Plus

Supplier: Anachemia
Danger

46564-320 46564-400
CA71007-532EA 2009.19 CAD
CA71007-532 CA71007-536
Hydrofluoric acid 47 - 51%, Environmental Grade Plus
Hydrofluoric acid
Formula: HF
Storage Temperature: Ambient
MDL Number: MFCD00011346
CAS Number: 7664-39-3
EINECS: 231-634-8
UN: 1790
ADR: 8,II

Order Now

Specification Test Results

Assay 47 - 51%
Aluminum (Al) 20 ppt max.
Antimony (Sb) 20 ppt max.
Arsenic (As) 50 ppt max.
Barium (Ba) 10 ppt max.
Beryllium (Be) 10 ppt max.
Bismuth (Bi) 10 ppt max.
Boron (B) 100 ppt max.
Cadmium (Cd) 10 ppt max.
Calcium (Ca) 10 ppt max.
Cerium (Ce) 10 ppt max.
Cesium (Cs) 10 ppt max.
Chromium (Cr) 10 ppt max.
Cobalt (Co) 10 ppt max.
Copper (Cu) 10 ppt max.
Dysprosium (Dy) 1 ppt max.
Erbium (Er) 1 ppt max.
Europium (Eu) 1 ppt max.
Gadolinium (Gd) 1 ppt max.
Gallium (Ga) 10 ppt max.
Germanium(Ge) 10 ppt max.
Gold(Au) 20 ppt max.
Hafnium (Hf) 10 ppt max.
Holmium (Ho) 1 ppt max.
Indium (In) 1 ppt max.
Iron (Fe) 10 ppt max.
Lanthanum (La) 10 ppt max.
Lead (Pb) 10 ppt max.
Lithium (Li) 10 ppt max.
Lutetium (Lu) 1 ppt max.
Magnesium (Mg) 10 ppt max.
Manganese (Mn) 10 ppt max.
Mercury (Hg) 50 ppt max.
Molybdenum (Mo) 10 ppt max.
Neodymium (Nd) 1 ppt max.
Nickel (Ni) 20 ppt max.
Niobium (Nb) 10 ppt max.
Palladium (Pd) 20 ppt max.
Platinum (Pt) 20 ppt max.
Potassium (K) 10 ppt max.
Praseodymium (Pr) 1 ppt max.
Rhenium (Re) 10 ppt max.
Rhodium(Rh) 20 ppt max.
Rubidium (Rb) 20 ppt max.
Ruthenium (Ru) 20 ppt max.
Samarium (Sm) 1 ppt max.
Scandium (Sc) 10 ppt max.
Selenium (Se) info only
Silver (Ag) 10 ppt max.
Sodium (Na) 10 ppt max.
Strontium (Sr) 10 ppt max.
Tantalum (Ta) info only
Tellurium (Te) 1 ppt max.
Terbium (Tb) 1 ppt max.
Thallium (Tl) 10 ppt max.
Thorium (Th) 1 ppt max.
Thulium (Tm) 1 ppt max.
Tin (Sn) 20 ppt max.
Titanium (Ti) 20 ppt max.
Tungsten (W) 20 ppt max.
Uranium (U) 1 ppt max.
Vanadium (V) 10 ppt max.
Ytterbium (Yb) 1 ppt max.
Yttrium (Y) 1 ppt max.
Zinc (Zn) 10 ppt max.
Zirconium (Zr) 10 ppt max.
Total sulfur 100 ppt max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR