Ethanol denatured ≥80% (denatured with methanol and ethyl acetate)

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Ethyl alcohol denatured

198V-24 198V-07
CA71007-234EA 3379.9 CAD
CA71007-234 CA71007-232
Ethanol denatured ≥80% (denatured with methanol and ethyl acetate)
Ethanol denatured

(DAG-2A)

Formula: H₃CCH₂OH
MW: 46.07 g/mol
MDL Number: MFCD00003568
CAS Number: 64-17-5
UN: 1986
ADR: 3 (6.1),II

Order Now

Specification Test Results

Ethanol 80.2% v/v min.
Methanol 12.95 - 14.45% v/v
Ethyl acetate 0.75 - 0.95% v/v
Density at 20°C 0.8106 g/ml max.
Colour (APHA) 5 max.
Acidity (as CH3COOH ) 0.003% w/v max.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR