Ethanol denatured, anhydrous

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

38332-540 38332-700 38332-549
CA71007-222CS 2563.88 CAD
CA71007-222 CA71007-224 CA71007-226 CA11011-114
Ethanol denatured, anhydrous
Ethanol denatured

(DAG-2A)

Formula: H₃CCH₂OH
MDL Number: MFCD00003568
CAS Number: 64-17-5
UN: 1170
ADR: 3,II

Order Now

Specification Test Results

Ethanol 84.46% v/v min.
Methanol 12.95 - 14.45% v/v
Water (H₂O) 0.14% v/v max.
Ethyl acetate 0.75 - 0.95% v/v
Density at 20°C 0.7922 g/l max.
Colour (APHA) 5 max.
Acidity (as CH₃COOH) 0.003% w/v max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR