Sodium dodecyl sulfate (SDS), Lab-Grade

Supplier: VWR
Danger

37122-300 37122-580
CA71007-096EA 414.31 CAD
CA71007-096 CA71007-098
Sodium dodecyl sulfate (SDS), Lab-Grade
Sodium dodecyl sulphate (SDS)
Formula: CH₃(CH₂)₁₁OSO₃Na
Melting Pt: 204…207 °C
Density: 1.1 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00036175
CAS Number: 151-21-3
EINECS: 205-788-1
UN: 1325
ADR: 4.1,II
Merck Index: 14,08636
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe4cb1741fba41f699a3be006d0b01b1b551898c&vid=c6df9a86fc351b319e68cc79c1c17f733b4fec67&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Anionic Surfactant (MW 289) 95.0 - 99.0%
Unsulfated Matter 1% max.
Sodium chloride (NaCl) 1% max.
Sodium sulfate 2.5% max.
Water (H2O) 1% max.
pH of a 1% solution 8.5 - 10.5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR