Dichloromethane ≥99.5% ACS

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

32667-540 32667-700 32667-950
CA71007-070EA 242.01 CAD
CA71007-070 CA71007-074 CA75812-788
Dichloromethane ≥99.5% ACS
Dichloromethane
Formula: CH₂Cl₂
Boiling Pt: 39.8 °C (1013 hPa)
Melting Pt: –95 °C
Density: 1.322 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000881
CAS Number: 75-09-2
EINECS: 200-838-9
UN: 1593
ADR: 6.1,III
Merck Index: 12,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bea1f8abe149a925f126e308d8aaee3a107495c9&vid=1a50c1b5a88f4f946c811d278fe40566f022efb4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (CH2Cl2) 99.5% min.
Appearance Clear
Colour (APHA) 10 max.
Residue after evaporation 0.002% max.
Titrable acid 0.0003 meq/g max.
Free halogens Passes Test
Water (H2O) 0.02% max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR