VWR® Buffer Solutions
Supplier: VWR
15209-700
15213-320
15213-360
15213-540
15213-700
15238-700
15206-700
15200-320
CA71006-712EA
42.17
CAD
CA71006-712
CA71006-718
CA71006-720
CA71006-724
CA71006-726
CA71006-728
CA71006-730
CA71006-734
CA71006-736
CA71006-740
CA71006-744
CA71006-756
CA71006-760
CA71006-762
CA71006-764
CA71006-766
CA71006-768
CA71006-772
CA71006-774
CA71006-776
CA71006-780
CA71006-784
CA71006-788
CA71006-798
CA71006-802
CA71006-804
CA71006-808
CA71006-812
CA71006-814
CA71006-816
CA71006-820
CA71006-708
CA71006-710
CA71006-840
CA71006-844
CA11020-850
CA11020-852
CA11020-924
CA11020-926
CA11021-928
CA11021-938
CA11021-948
CA11021-952
VWR® Buffer Solutions
Buffers
pH Calibration Buffers
These buffer solutions are for calibrating pH meters
They are traceable to NIST standards.