Barium chloride dihydrate ≥99.0% ACS

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Barium dichloride

176V-21 176V-22
CA71006-648EA 2474.99 CAD
CA71006-648 CA71006-646
Barium chloride dihydrate ≥99.0% ACS
Barium chloride dihydrate
Formula: BaCl₂·2H₂O
MW: 244.27 g/mol
Boiling Pt: 1560 °C (1013 hPa)
Melting Pt: 960 °C
Density: 3.86 g/cm³ (25 °C)
MDL Number: MFCD00149154
CAS Number: 10326-27-9
UN: 1564
ADR: 6.1,III
Merck Index: 14,00971

Order Now

Specification Test Results

Assay (BaCl2.2H2O) 99.0% min.
Loss on drying at 150°C 14.0 - 16.0 %
pH of a 5% Solution at 25°C 5.2 - 8.2
Insoluble Matter 0.005% max.
Oxidizing substances (as NO3) 0.005% max.
Calcium (Ca) 0.05% max.
Potassium (K) 0.0025% max.
Sodium (Na) 0.005% max.
Strontium (Sr) 0.1% max.
Heavy metals (as Pb) 5 ppm max.
Iron (Fe) 2 ppm max.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR