Ammonia solution 20 - 22%, Environmental Grade VWR®

Supplier: Anachemia
Danger

05560-320 05560-540
CA71006-570EA 303.15 CAD
CA71006-570 CA71006-573
Ammonia solution 20 - 22%, Environmental Grade VWR®
Ammonia solution
Formula: NH₃
Density: 0.92 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00066650
CAS Number: 1336-21-6
EINECS: 215-647-6
UN: 2672
ADR: 8,III

Order Now

Specification Test Results

Assay (as NH3) 20 - 22%
Colour (APHA) 10 max.
Aluminum (Al) 1 ppb max.
Antimony (Sb) 0.5 ppb max.
Arsenic (As) 1 ppb max.
Barium(Ba) 0.1 ppb max.
Beryllium (Be) 0.1 ppb max.
Bismuth (Bi) 0.1 ppb max.
Cadmium (Cd) 0.5 ppb max.
Calcium (Ca) 1 ppb max.
Cerium (Ce) 0.1 ppb max.
Cesium (Cs) 0.1 ppb max.
Chromium (Cr) 0.5 ppb max.
Cobalt (Co) 0.5 ppb max.
Copper (Cu) 0.5 ppb max.
Dysprosium (Dy) 0.1 ppb max.
Erbium (Er) 0.1 ppb max.
Europium (Eu) 0.1 ppb max.
Gadolinium (Gd) 0.1 ppb max.
Gallium(Ga) 0.1 ppb max.
Germanium(Ge) 0.1 ppb max.
Gold(Au) 0.5 ppb max.
Hafnium (Hf) info only
Holmium (Ho) 0.1 ppb max.
Indium (In) 0.1 ppb max.
Iron (Fe) 1 ppb max.
Lanthanum (La) 0.1 ppb max.
Lead (Pb) 0.1 ppb max.
Lithium (Li) 0.1 ppb max.
Lutetium (Lu) 0.1 ppb max.
Magnesium (Mg) 1 ppb max.
Manganese (Mn) 0.5 ppb max.
Mercury (Hg) 0.2 ppb max.
Molybdenum (Mo) 0.5 ppb max.
Neodymium (Nd) 0.1 ppb max.
Nickel (Ni) 0.5 ppb max.
Niobium (Nb) 0.1 ppb max.
Palladium (Pd) info only
Platinum (Pt) info only
Potassium (K) 1 ppb max.
Praseodymium (Pr) 0.1 ppb max.
Rhenium (Re) info only
Rhodium(Rh) 0.5 ppb max.
Rubidium (Rb) 0.1 ppb max.
Ruthenium (Ru) info only
Samarium (Sm) 0.1 ppb max.
Scandium (Sc) 0.1 ppb max.
Selenium(Se) 1 ppb max.
Silver (Ag) 0.5 ppb max.
Sodium (Na) 1 ppb max.
Strontium (Sr) 0.1 ppb max.
Tellurium (Te) 0.1 ppb max.
Terbium (Tb) 0.1 ppb max.
Thallium (Tl) 0.1 ppb max.
Thorium (Th) 0.1 ppb max.
Thulium (Tm) 0.1 ppb max.
Tin (Sn) 0.5 ppb max.
Titanium (Ti) 0.5 ppb max.
Tungsten (W) 0.1 ppb max.
Uranium (U) 0.1 ppb max.
Vanadium (V) 0.5 ppb max.
Ytterbium (Yb) 0.1 ppb max.
Yttrium (Y) 0.1 ppb max.
Zinc (Zn) 0.5 ppb max.
Zirconium (Zr) 0.1 ppb max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR