Ammonium chloride ACS
Supplier: VWR
|
Warning
|
05106-300
05106-380
CA71006-562EA
581.91
CAD
CA71006-562
CA71006-564
CA71006-566
Ammonium chloride ACS
Ammonium chloride
Formula:
NH₄Cl Melting Pt: 335 °C Density: 1.5256 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011420 CAS Number: 12125-02-9 EINECS: 235-186-4 Merck Index: 13,00510 |
|
Specification Test Results
Assay (NH4Cl) | 99.5% min. |
pH of a 5% Solution at 25°C | 4.5 - 5.5 |
Insoluble Matter | 0.005% max. |
Residue after ignition | 0.01% max. |
Calcium (Ca) | 0.001% max. |
Magnesium (Mg) | 5 ppm max. |
Heavy metals (as Pb) | 5 ppm max. |
Iron (Fe) | 2 ppm max. |
Phosphate (PO4) | 2 ppm max. |
Sulfate (SO4) | 0.002% max. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...