Ammonium acetate ≥97%, crystals
Supplier: VWR
Synonyms:
Acetic acid ammonium salt
ANAC04566-820
CA71006-554EA
8683.02
CAD
CA71006-554
Ammonium acetate ≥97%, crystals
Ammonium acetate
Formula:
H₃CCOONH₄ MW: 77.08 g/mol Melting Pt: 114 °C Density: 1.17 g/cm³ (20 °C) Flash Pt: 136 °C |
MDL Number:
MFCD00013066 CAS Number: 631-61-8 Merck Index: 13,00495 |
Specification Test Results
Assay | 97% min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...