Acetone ≥99.5% ACS

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: 2-PropanoneDMKPropanoneDimethyl ketone

055V-03 ANAC00884-700 055V-33 169V-03 055V-07 169V-07
CA71006-468CS 847.84 CAD
CA71006-468 CA71006-466 CA71006-476 CA71006-464 71006-476 CA71006-474
Acetone ≥99.5% ACS
Acetone
Formula: CH₃COCH₃
MW: 58.08 g/mol
Boiling Pt: 56.2 °C (1013 hPa)
Melting Pt: –95.4 °C
Density: 0.792 g/cm³ (20 °C)
Flash Pt: <–20 °C (closed cup)
MDL Number: MFCD00008765
CAS Number: 67-64-1
UN: 1090
ADR: 3,II
Merck Index: 13,00067
https://ca.vwr.com/stibo/web/std.lang.all/06/07/720607.jpg

Order Now

Specification Test Results

Assay (CH3)2CO 99.5% min.
Color (APHA) 10 max.
Residue after evaporation 0.001% max.
Solubility in water Passes Test
Titrable acid 0.0003 meq/gmax.
Titrable base 0.0006 meq/g max.
Aldehyde (as HCHO) 0.002% max.
Isopropyl Alcohol 0.05% max.
Methanol 0.05% max.
Substances reducing KMnO4 Passes Test
Water (H2O) 0.5% max.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR