Acetone, Accusolv™, distilled
Supplier: Anachemia
Synonyms:
2-Propanone
, DMK
, Propanone
, Dimethyl ketone
ANAC00870-540
ANAC00870-540
CA71006-454CS
1066.75
CAD
CA71006-454
71006-454
Acetone, Accusolv™, distilled
Acetone
Suitable for Spectrophotometry, HPLC and Pesticide Residue Analysis.
Filtered through a 0.2µm filter
Formula:
CH₃COCH₃ MW: 58.08 g/mol Boiling Pt: 56.2 °C (1013 hPa) Melting Pt: –95.4 °C Density: 0.792 g/cm³ (20 °C) Flash Pt: <–20 °C (closed cup) |
MDL Number:
MFCD00008765 CAS Number: 67-64-1 UN: 1090 ADR: 3,II Merck Index: 13,00067 |
Specification Test Results
UV absorbance (1 cm cell vs H2O) 330 nm | 1.000 max. |
UV absorbance (1 cm cell vs H2O) 340 nm | 0.060max. |
UV absorbance (1 cm cell vs H2O) 350 nm | 0.010 max. |
UV absorbance (1 cm cell vs H2O) 375 nm | 0.005 max. |
UV absorbance (1 cm cell vs H2O) 400 nm | 0.005max. |
Assay (GC) not corrected for water | 99.9% min. |
Water (H2O) | 0.50% max. |
Titrable acid | 0.0003 meq/g max. |
Titrable base | 0.0006 meq/g max. |
Residue after evaporation | 1 mg/l max. |
Electron capture GC as Heptachlorepoxide | 10 ng/l max. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...