Eppendorf® Easypet® 3 Electronic Pipette Controller
Supplier: Eppendorf
4430000018 4430000018
CA11027-980EA 1117.45 CAD
CA11027-980 10860-782
Eppendorf® Easypet® 3 Electronic Pipette Controller
Pipettes Pipette Controllers
Electronic pipetting doesn't come any more advanced than the Easypet 3 from Eppendorf. Designed to work with Eppendorf serological pipettes, you get complete speed control and utmost precision in an extremely light and well-balanced body that fits comfortably in your hand, for fatigue-free pipetting.
  • Intuitive and convenient speed adjustment simply done with the tips of your fingers
  • Lightweight, well-balanced and ergonomic design that allows for fatigue-free pipetting
  • Vibrant backlit LEDs provide optical feedback of the remaining battery life
  • Lithium
More Product Information