Desiccator, Polycarbonate, Chemglass
Supplier: Chemglass
CG-1226-10 CG-1226-11 CG-1226-12
80060-658EA 231.42 CAD
80060-658 80060-660 80060-662
Desiccator, Polycarbonate, Chemglass
Desiccators Desiccators, Plastic
Clear polycarbonate desiccator with extra-heavy walls to eliminate the danger of implosion
Desiccator will hold a vacuum for a period of 24 hours. Space saving design of the desiccator dome provides maximum interior clearances, giving an average of 13% greater interior volume than hemispherical domes. The supplied polypropylene stopcock has a PTFE plug which turns easily, allowing a slow, gentle entrance of air to prevent unnecessary turbulence and crucible damage. Stopcock port accepts 1/4” I.D. tubing. Supplied complete with a neoprene o-ring unaffected by temperatures down to 0°C.More Product Information