Corning® Pasteur Pipettes
Supplier: Corning
7095B-NMR 7095D-5X 7095D-9
14673-156CS 264.73 CAD
14673-156 36360-536 89090-042 89090-044 52947-802
Corning® Pasteur Pipettes
Pipettes Pasteur/Transfer Pipettes
Soda-lime or borosilicate glass.
  • Single use eliminates the danger of cross-contamination of valuable specimens or laboratory reagents
  • Features a constriction one inch below the top to facilitate plugging
  • Available in convenient bulk packs
These disposable Pasteur pipettes are ideal for rapid non volumetric transfer work in bacteriology, immunology, hematology and serology studies, as well as blood bank and clinical chemistry procedures.