CTA (Cystine Tryptic Agar), Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
Y12 Y13 Y15 Y17 Y20 Y24
CA89492-726PK 586.14 CAD
CA89492-726 CA89492-728 CA89492-730 CA89492-732 CA89492-734 CA89492-736 CA76047-226 CA76047-228
CTA (Cystine Tryptic Agar), Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CTA (Cystine Tryptic Agar), 3mL Deep, 13x100mm tube, optically clear, shatter resistant, polycarbonate tube, order by the package of 20, by Hardy Diagnostics.
Hardy Diagnostics CTA Media are recommended for the determination of carbohydrate fermentation by fastidious organisms such as Neisseria spp. They are also used for the detection of bacterial motility and serve as a culture media for microorganism maintenance.
Order Now
SPECIFICATIONS
Environmentally Preferable
Storage temperature 2-8C