Sabdex (Sabouraud Dextrose) Agar, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
W72 U445
CA52428-858PK 65.79 CAD
CA52428-858 CA76809-014
Sabdex (Sabouraud Dextrose) Agar, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Sabdex (Sabouraud Dextrose) Agar with Chloramphenicol, deep fill, 15x100mm plate, order by the package of 10, by Hardy Diagnostics
(Sabourauds Dextrose Agar with Chloramphenicol) For the selective isolation of fungi.