SP4 Broth, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
R84 R85 R86 R87
CA89497-628PK 321.9 CAD
CA89497-628 CA89498-940 CA89498-942 CA89498-944
SP4 Broth, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
SP4 Broth,for Mycoplasma and Ureaplasma, 2ml, 13x100mm tube, optically clear, shatter resistant, polycarbonate tube, order by the package of 20, by Hardy Diagnostics.
SP4 Broths 89498-940, 89498-942, and 89498-944 sold seperatly.
Ordering information: SP4 Broths 89498-940, 89498-942, and 89498-944 sold seperatly.
Order Now
SPECIFICATIONS
Environmentally Preferable
Storage temperature 2-8C