BCSA, Selective for Burkholderia cepacia
Supplier: HARDY DIAGNOSTICS CA
G09 GA09
CA89423-720PK 194.77 CAD
CA89423-720 CA76288-334
BCSA, Selective for Burkholderia cepacia
Media for Microbiology Dehydrated Media for Microbiology
Burkholderia cepacia Selective Agar is ideal for the selective isolation and differentiation of Burkholderia (Pseudomonas) cepacia from sputum samples from cystic fibrosis patients and other clinical specimens.
Packaging: Pack of 10.
Order Now
SPECIFICATIONS
Environmentally Preferable
Storage temperature 2...8 °C