LabLink Power Strips, PG LifeLink
Supplier: PG Lifelink
VPS130-18 VPS130-24 VPS130-36 VPS130-48 VPS130-60
89422-952EA 531.43 CAD
89422-952 89422-954 89422-956 89422-958 89422-960
LabLink Power Strips, PG LifeLink
LabLink Power Strips are made of high quality extruded aluminum and can be permanently mounted to your work station.
  • Extruded from #6063-T5 Aluminum Alloy
  • Dimensions 1³/₄×2³/₄×18" - 60"
  • Ideal for Mobile Benches
  • Single Circuit Configuration
  • Clear Satin Anodized Finish is Durable and Corrosion Resistant
  • 20 Amp (NEMA 5-20R) Grey Duplex Receptacles
  • 8 Foot, 3 Conductor, Black Power Cord with Straight Blade Plug (NEMA5-20P)
Power strips are available in lengths of 18", 24", 36", 48" and 60", providing more functionality for today's modular lab environment. Cover with 2 end plates.
Certifications: C-UL Listed.
Ordering information: Not to be used in conjunction with an extension cord or another power strip.