LabLink Surface Mount Raceway, Versa-Duct, PG LifeLink
Supplier: PG Lifelink
VRW130BC-48 VRW130BC-60 VRW130BC-72 VRW130BC-84 VRW130BC-96 VRW130BC-108 VRW130BC-120 VRW252BC-48 VRW252BC-60 VRW252BC-72 VRW252BC-84 VRW252BC-96 VRW252BC-108 VRW252BC-120
89422-924EA 395.88 CAD
89422-924 89422-926 89422-928 89422-930 89422-932 89422-934 89422-936 89422-938 89422-940 89422-942 89422-944 89422-946 89422-948 89422-950
LabLink Surface Mount Raceway, Versa-Duct, PG LifeLink
Electrical Sockets
LabLink Surface Mount Raceway (Versa-Duct), made from anodized extruded aluminum, offers a superior design to combine electrical and/or data into one unified system.
  • Extruded from #6063-T5 Aluminum Alloy
  • Single Channel Assembly (1³/₄×2³/₄×48 – 120")
  • Double Channel Assembly (2¹/₄×5¹/₄×48 – 120")
  • Single Circuit Configuration
  • 12" Pigtails are provided for Electrical Connections
  • Clear Satin Anodized Finish is Durable and Corrosion Resistant
  • 12" Removable Cover Plates for Easy Access in the Field
  • 20 Amp (NEMA-5-20R) Grey Duplex Receptacles
  • Grey Communication
More Product Information