Sodium ethoxide 20% in ethanol for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Sodium ethylate, Sodium ethanolate

M-Clarity™ quality level = MQ200

8.18199.0100 8.18199.1000 8.18199.9050 8.18199.1000 8.18199.9050
CA8.18199.0100EA 48 CAD
CA8.18199.0100 EM8.18199.1000 EM8.18199.9050 CA8.18199.1000 CA8.18199.9050
Sodium ethoxide 20% in ethanol for synthesis, Sigma-Aldrich®
Sodium ethoxide
Formula: C₂H₅NaO
Boiling Pt: 91 °C (1013 hPa)
Density: 0.88 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012417
CAS Number: 141-52-6
UN: 3274
ADR: 3.8,II

Order Now

Specification Test Results

Assay (acidimetric) 20.0 - 22.0 %
Density (d 20 °C/ 4 °C) 0.875 - 0.880


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR