DL-Limonene mixture of D- and L-form (∼1:1) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: (±)-4-Isopropenyl-1-methyl-1-cyclohexene(±)-p-Mentha-1,8-diene(±)-Dipentene(±)-LimoneneDipentene(±)-1-Methyl-4-(1-methylvinyl)cyclohexene

M-Clarity™ quality level = MQ200

8.14546.1000 8.14546.1000
EM8.14546.1000CS 965.05 CAD
EM8.14546.1000 CA8.14546.1000
DL-Limonene mixture of D- and L-form (∼1:1) for synthesis, Sigma-Aldrich®
DL-Limonene
Formula: C₁₀H₁₆
MW: 136.24 g/mol
MDL Number: MFCD00062992
CAS Number: 138-86-3
UN: 2052
ADR: 3,III

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point -89 °C
Molar Mass 136.24 g/mol
Boiling Point 178 °C (1013 hPa)
Vapor Pressure 2.1 hPa (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR