Benzoin for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: 2-Hydroxy-2-phenylacetophenone

M-Clarity™ quality level = MQ200

8.01776.0250 8.01776.1000 8.01776.0250 8.01776.1000
EM8.01776.0250CS 1761.7 CAD
EM8.01776.0250 EM8.01776.1000 CA8.01776.0250 CA8.01776.1000
Benzoin for synthesis, Sigma-Aldrich®
Benzoin
Formula: C₁₄H₁₂O₂
MW: 212.25 g/mol
MDL Number: MFCD00004496
CAS Number: 119-53-9
EINECS: 204-331-3

Order Now

Specification Test Results

Solubility 0.3 g/l (25 °C)
Melting Point 132 - 134 °C
Molar Mass 212.24 g/mol
Bulk Density 350 kg/m3
Boiling Point 344 °C (1023 hPa)
Vapor Pressure 1.3 hPa (136 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR