SP Bel-Art Screw Cap Jars, Polypropylene, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
H17910-0000 H17911-0000
70680-512PK 34.65 CAD
70680-512 70680-514 70680-518 70680-520
SP Bel-Art Screw Cap Jars, Polypropylene, Bel-Art Products, a part of SP
Jars General Use Jars
White polypropylene jars and screw caps in an assortment of sizes are FDA approved and are excellent for storage of liquid or dry stock, or for use with samples.
  • White polypropylene construction
  • Suited for liquid or dry stock storage
  • Autoclavable
Jars are autoclavable at 121˚C (250˚F). Screw caps, with PE foam lining, are not autoclavable.