Disposable Serological Pipettes, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
72120-1110 72120-11100 72120-21100 72120-5110 72120-10110
14673-200CS 1294.8 CAD
14673-200 14673-202 14673-204 14673-206 14673-208
Disposable Serological Pipettes, Kimble Chase
Pipettes Serological Pipettes
Pipettes are non-plugged and nonsterile, with a uniform pulled tip.
Calibrated 'to deliver' to the tip, with two rings near the top denoting that the small amount remaining in the tip after free delivery has ceased must be blown out and added to the main volume delivered. Pipettes are provided with negative graduations to increase versatility. Made from borosilicate glass in compliance with ASTM Specification E438, Type I, Class B glass requirements. Bulk packed in sealed plastic bags and overpacked in shelf packs. Nonsterile.