Model CN-DT Total Chlorine Test Kit, Hach
Supplier: Hach
2471100 2471100
CA24711-00EA 833.45 CAD
CA24711-00 89500-736 CA89500-736
Model CN-DT Total Chlorine Test Kit, Hach
Environmental Test Kits
Chlorine is important for sanitizing swimming pools, cooling towers, other industrial equipment, and in the treatment of municipal wastewater. Its measurement and control are vital for both safety and economic reasons. This kit utilizes the Hach Digital Titrator and contains 100 tests.
Hach Digital Titrator offers higher levels of precision and accuracy than drop count procedures, with reproducibility comparable to a buret titration
Items with this mark may be considered hazardous under some shipping conditions.If necessary, we will change your selected shipping method to accomodate these items.
Rugged titration device is ideal for field use, with no glass burets required. Kit contains all required reagents and apparatus to perform analysis in the field.
Ordering information: This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States. This kit includes four reagents, flask, digital titrator, delivery tube, cylinder, bottle, clippers, instruction sheet, and carrying case.
Order Now
SPECIFICATIONS
Environmentally Preferable
Method Digital Titrator/Thiosulfate