National™ Micro-Centrifugal Filters, Nonsterile, Thermo Scientific
Supplier: Thermo Scientific
F2517-1 F2517-2 F2517-3 F2517-4 F2517-5 F2517-6 F2517-7 F2517-8 F2517-10 F2517-9
66064-442PK 652.02 CAD
66064-442 66064-444 66064-446 66064-448 66064-450 66064-452 66064-454 66064-456 97006-920 97006-922
National™ Micro-Centrifugal Filters, Nonsterile, Thermo Scientific
Filters Centrifugal Filters
Filter volumes as low as 50 µl with low hold-up volume with these National™ micro-centrifugal filters.
  • 10000×g maximum centrifugal force
  • Non sterile
  • Bulk packed
The filters feature a virgin polypropylene filter housing with tapered 2 ml, capped receiver tube, and can be used with any benchtop microcentrifuge.