VWR® Traceable® Platinum High-Accuracy Refrigerator Thermometer
Supplier: VWR International
89369-146 89369-148
89369-146EA 426.87 CAD
89369-146 89369-148
VWR® Traceable® Platinum High-Accuracy Refrigerator Thermometer
Thermometers Digital Thermometers
Therometer displays minimum/maximum temperatures for last 24 hours for each hour and previous 7 days for each day, or last 30 days for every day.
  • Ideal for refrigerators
  • Displays exact time and date for all readings
  • Highly accurate across entire temperature range
  • Visual LED and audible alarm
  • Stainless steel probe
Time/date stamp key shows the exact time and date for all minimum and maximum readings
Triple display shows current minimum/maximum temperatures. Alarm is settable in 0.1° increments and signals when temperature rises above or fall belowMore Product Information