VWR® Traceable® Memory-Card Refrigerator/Freezer Thermometer
Supplier: VWR International
89369-154 89369-156
89369-154EA 797.74 CAD
89369-154 89369-156
VWR® Traceable® Memory-Card Refrigerator/Freezer Thermometer
Thermometers Digital Thermometers
Traceable® Memory-Card Refrigerator/Freezer Thermometer is a stand-alone unit with data logging capabilities.
  • Insulated from rapid temperature changes
  • Does not disturb seal of closed doors
  • Unique 4-line display
  • Captures a continuous stream of data
  • Records up to 5.9 million readings
It records readings from once a minute up to once every 720 minutes on a removable standard SD memory card
Sealed bottle sensor is insulated from rapid temperature changes when refrigerator door is opened. Solution in sealed bottle is generally recognized as safe by the FDA. The cable is easy for the refrigerator door to close and seal. Minimum/Maximum feature displays highest/lowest readings for any time period. Alarm sound and flashes a bright red LED when temperature rises above or falls below user-set trip points, alarm sets is 0.1° increments. Unique 4-line display shows temperature of probe, ambient temperature, humidity and time of day.
Thermometers captures 5.9 million readings with the supplied 256 mb removable SD memory card. Memory card reader plugs into any USB computer port for an instant report. Text report includes date or time-of-day and sensor readings, it may be sent to any spreadsheet or database. Capturing a continuous stream of data shows the behavior of refrigerators, room areas or tests. The bottle probe model is ideal for monitoring refrigerators, freezers and incubators; the stainless steel probe model is ideal for QC and research. Bottle probe meets all CDC, federal and state refrigerator thermometer requirements. Both models include internal sensors to measure the ambient temperature and humidity - ideal for monitoring store rooms, labs and critical areas.
Certifications: To assure accuracy, an individually serial-numbered Traceable® Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA. It indicates traceability to standards provided by NIST.
Ordering information: Supplied with probe, cable, 256 mb SD memory card, USB memory card reader, magnetic mounting strips, Velcro®, wall mount, bench stand, batteries, adaptor, Traceable® certificate.