Filter Adapters, Neoprene, Chemglass
Supplier: Chemglass
CG-1401-10 CG-1401-11 CG-1401-12 CG-1401-13 CG-1401-14 CG-1401-15 CG-1401-16
80061-058EA 13.7 CAD
80061-058 80061-060 80061-062 80061-064 80061-066 80061-068 80061-070
Filter Adapters, Neoprene, Chemglass
Supports Support Sleeves
Neoprene filter adapters are designed to fit between the filtering funnel and flask.
For use with Buchner funnels, Grooch crucibles, filter tubes, etc. Can be used individually or nesting with adjacent sizes. Eliminates the boring of rubber stoppers and the difficulties of inserting and removing glass stems in stoppers. Neoprene adapters withstand autoclaving.