Stir-Lube, Ace Glass
Supplier: Ace Glass
8117-10 8117-20
89062-224EA 59.33 CAD
89062-224 89062-226
Stir-Lube, Ace Glass
Lubricants
A superior, low melting, silicone-based lubricant which liquifies at body temperature
Due to its composition, only a very thin film of Stir-Lube needs to be applied to a stirring shaft to increase bearing and shaft life at least three times over that of a bearing lubricated with glycerine. Non-cooled ACE bearing can be operated at 1500rpm and water-cooled bearings up to 2000rpm for many hours with negligible wear.