ULTRA-WARE® Conical Bottom HPLC Reservoir, Plastic-Coated, Kimble Chase
Supplier: Kimble Chase
953901-2002 953901-5002 953922-0252 953922-0502 953922-1002 953922-1003 953922-2002 953922-5002 953901-0252 953901-0502 953901-1002 953901-1003
KT953901-2002EA 814.23 CAD
KT953901-2002 KT953901-5002 KT953922-0252 KT953922-0502 KT953922-1002 KT953922-1003 KT953922-2002 KT953922-5002 KT953901-0252 KT953901-0502 KT953901-1002 KT953901-1003 KT953901-2003
ULTRA-WARE® Conical Bottom HPLC Reservoir, Plastic-Coated, Kimble Chase
Bottles HPLC Bottles
ULTRA-WARE® Reservoirs have been specially designed for the preparation, storage and delivery of all liquid chromatography mobile phases.
ULTRA-WARE Reservoirs are recommended for use under vacuum for filtration and vacuum degassing of mobile phase solvents. ULTRA-WARE Reservoirs can also be used at internal pressures up to 6 psig for helium sparging and blanketing of the mobile phase. The conical bottom reservoir delivers virtually all of the mobile phase without dangerous reservoir tilting.
Each ULTRA-WARE Reservoir is manufactured from the most chemically inert glass available toMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable