SteriPettes, Kimble Chase, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
63A55 63A55P 63B95 63B95P
89218-062CS 173.01 CAD
89218-062 89218-064 89218-066 89218-068
SteriPettes, Kimble Chase, DWK Life Sciences
Pipettes Pasteur/Transfer Pipettes
SteriPettes are sterile, disposable Pasteur pipettes produced from soda-lime, gamma-irradiated glass.
These pipettes are either non-plugged or cotton-plugged.