Fetal bovine serum (FBS), premium grade, Corning®
Supplier: Corning
35-015-CV 35-016-CV
CA45001-106EA 1182.12 CAD
CA45001-106 CA45001-108
Fetal bovine serum (FBS), premium grade, Corning®
Sera (Serum)
FBS is a commonly used cell culture media supplement which helps sustain cell growth and satisfy metabolic requirements.
  • Sera is aseptically collected and processed in a cGMP facility
  • Each batch of raw serum is screened for endotoxin, residual hemoglobin concentration, and microbiological quality before being used in production
  • Batches of raw serum are pooled and continuously mixed prior to filtration to ensure consistent quality throughout the entire lot
  • Final lots are tested for endotoxin, hemoglobin, pH, mycoplasma, and growth promotion. Biochemical assays and virus testing are also available upon request
  • Premium FBS is sourced from U.S. herds
  • Heat-inactivated Fetal Bovine Serum (FBS) is heated at 56°C for 30 minutes