myCart™ Series Utility Carts, Metro™
Supplier: Metropolitan Wire
MY1627-24G MY1627-34G MY2030-24G MY2030-34G MY2636-25G MY2636-35G MY1627-24BL MY1627-34BL MY2030-24BL MY2030-34BL MY2636-25BL MY2636-35BL MY1627-34BU MY2030-24BU MY2030-34BU MY2636-25BU MY2636-35BU MY1627-24G MY1627-34G MY2030-24G MY2030-34G MY2636-25G MY2636-35G MY1627-24BL MY1627-34BL MY2030-24BL MY2030-34BL MY2636-25BL MY2636-35BL MY1627-34BU
89501-326EA 707.7 CAD
89501-326 89501-328 89501-330 89501-332 89501-334 89501-336 89501-338 89501-340 89501-342 89501-344 89501-346 89501-348 89501-352 89501-354 89501-356 89501-358 89501-360 CA89501-326 CA89501-328 CA89501-330 CA89501-332 CA89501-334 CA89501-336 CA89501-338 CA89501-340 CA89501-342 CA89501-344 CA89501-346 CA89501-348 CA89501-350 CA89501-352 CA89501-354 CA89501-356 CA89501-358 CA89501-360
myCart™ Series Utility Carts, Metro™
Trolleys
Utility carts feature injection molded polyethylene shelves for maximum corrosion resistance and durability.
  • Built for use in corrosivie environments
  • Carts feature smooth shelf surfaces for easy cleaning
  • Ship's edge retains small spills and prevents items from sliding during transport
  • Shelves will not chip, dent or peel
New features include easy grip handle with integrated utility tray. MY2636 Series includes slot in handle to hold scanners and labelers. Inset design of lower shelf provides 50% more legroom than conventional carts. You won't hit your shins or kick the cart. Special label included to help you identify this as myCart!
Upgrade models available with stainless steel posts for added corrosion resistance.
Certifications: NSF Listed.
Ordering information: Available in a choice of three colors, black, gray or blue. All blue carts feature built in Microban® antimicrobial product protection which inhibits the growth of bacteria, mold, mildew and fungi that cause orders, stains and product degredation. All carts are shipped knocked down for easy assembly. Stated shelf and cart capacities are for weight evenly distributed.