VWR® Basic Chromatography Refrigerators with Glass Doors and Natural Refrigerant
Supplier: VWR International
10791-614 10791-616 10791-622 10791-628 10018-856 10018-858 10018-862 GDM72WHTSSPDM 75794-802 75794-804 75840-438 75840-440
10791-614EA 12188.88 CAD
10791-614 10791-616 10791-622 10791-628 10018-856 10018-858 10018-862 10018-868 75794-802 75794-804 75840-438 75840-440
VWR® Basic Chromatography Refrigerators with Glass Doors and Natural Refrigerant
Refrigerators
Swing door, upright laboratory chromatography refrigerators maintain an average temperature of 2 to 8 °C (35.6 to 46.4 °F)
  • Units maintain an average temperature of 2 to 8 °C (35.6 to 46.4 °F)
  • Equipped with energy efficient thermal insulated glass doors
  • Three-year parts and labor warranty
  • Additional two-year warranty on compressor parts
Units are equipped with low E, double-pane thermal insulated glass doors for energy efficiency
All units feature a natural refrigerant that is environmentally friendly. The R290 hydrocarbon refrigerant emits zero (0) ozone depletion potential (ODP), and three (3) global warming potential (GWP).
Units are supplied standard with the LAE temperature control, which offers exterior digital display and alarm function.
The 23 cu.ft. units equipped with half shelves feature one access port, one duplex outlet, and one chromatography mast. The 23 cu.ft. units equipped with full shelves feature one access port and one duplex outlet. The 41, 47, 49 and 72 cu.ft. units feature two access ports, one duplex outlet, and one chromatography mast.
All refrigerators are supplied standard with adjustable, heavy-duty, PVC-coated white wire shelves and LED lighting.
Certifications: C-UL US listed. NSF certified. Natural Refrigerant.
Ordering information: Additional full and half white wire shelves can be ordered seperately for any VWR® Single Glass Door Chromatography Refrigerators. Models with swing doors are available on request, please contact your local Avantor sales office for details. Optional Chart Recorder, Alarm, and Thermometer accessories require factory installation. Refrigerators come with a three-year parts and labor warranty, with an additional two-year warranty on the compressors.