Q500 Sonicator, Qsonica
Supplier: Qsonica
Q500A-110 Q500-220 Q500A-220 Q500-110
89207-040EA 11878.71 CAD
89207-040 89207-038 89207-036 89207-042
Q500 Sonicator, Qsonica
Homogenizers Ultrasonic Cell Disruptors
The Q500 is a powerful ultrasonic processor featuring programmable operation and a digital display of operating parameters
Adjustable pulse On and Off times can be programmed from 1 second to 1 minute. Total programming has a maximum setting of 10 hours. A wide variety of probes and accessories are available to handle virtually any application. Lead free components.
Popular applications include nanoparticle dispersion, creating emulsions, cell lysis and homogenization.
Accessories information: Horn extenders, also available in solid or tapped varieties, can reach into long neck vessels. Half-wave extenders add 12.7 cm (5") to the horn’s length; full-wave extenders add 25.4 cm (10") to the horn’s length.