Micro Capillary Tubes, Disposable, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
71900-10 71900-100 71900-20 71900-25
72690-002CS 669.43 CAD
72690-002 72690-004 72690-006 72690-008
Micro Capillary Tubes, Disposable, Kimble Chase
Pipettes Microcapillaries
Capillary action draws the fluid into the tube. To dispense, simply cover the hole on the rubber bulb and squeeze. Pipettes are 5" long with a single black capacity ring. They are color-coded for selection of the correct size.
  • Packaged in easy-open cylinders for convenient dispensing
  • Clean and ready to use
  • Filling is easy
Certifications: Micro Capillary pipettes made of soda lime glass in compliance with ASTM Specification E438, Type II glass requirements.
Packaging: Packaging is designed from ASTM Specification E672 requirements. Each cylinder contains 250 pipettes and one pipette device.