D.O. Probe Service Kit, Hach
Supplier: Hach
5196800 51968-00
CA51968-00EA 484.92 CAD
CA51968-00 97021-610
D.O. Probe Service Kit, Hach
Electrolytes Electrode Filling Solutions
DO Probe Service Kit comes with two membranes, electrolyte filling solution and cloth.