AlphaTec® 58-335, Nitrile Gloves, with AQUADRI® Technology, Ansell
Supplier: Ansell Canada
114019 114020 114021 114022 114023
CA89413-064CS 1502.39 CAD
CA89413-064 CA89413-066 CA89413-068 CA89413-070 CA89413-072
AlphaTec® 58-335, Nitrile Gloves, with AQUADRI® Technology, Ansell
Gloves
Ansell’s new, proprietary AQUADRI® technology delivers the properties of a sponge with great moisture absorption capacity, featuring AquaDri® Nitrile Foam Lining, thus keeping hands drier for a longer period of time resulting in improved comfort and performance.
  • Superior puncture and abrasion protection
  • Improved dry and wet grip
  • Gauntlet cuffs prevent dripping onto forearms
  • Great moisture absorption capacity
These gloves are designed to deliver optimal results in wet or dry work environments where chemical protection is crucial.
Ideal uses areMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Ambidextrous/Hand-specific Hand-specific
Finish Diamond Palm