AlphaTec® 58-330, Nitrile Gloves, with AQUADRI® Technology, Ansell
Supplier: Ansell Canada
114014 114015 114016 114017 114018
CA89413-054CS 1373.44 CAD
CA89413-054 CA89413-056 CA89413-058 CA89413-060 CA89413-062
AlphaTec® 58-330, Nitrile Gloves, with AQUADRI® Technology, Ansell
Gloves
Ansell proprietary AQUADRI® technology delivers the properties of a sponge with great moisture absorption capacity, featuring AquaDri® Nitrile Foam Lining, thus keeping hands drier for a longer period of time resulting in improved comfort and performance.
  • Optimal performance in wet or dry environments
  • Resistant to abrasion damage
  • Gauntlet cuffs
These gloves are designed to deliver optimal results in wet or dry work environments where chemical protection is crucial.
Ideal uses are changing oil, fixing pumps, chemical mixing/handling, cleaning, opening/draining pumps and valves, painting, applying sealants and sampling/testing.
The high-performance nitrile compound offers superior snag, puncture and abrasion protection over rubber or neoprene gloves. The diamond palm grip provides improved dry and wet grip while the foldable gauntlet cuff gutter prevents dripping on the forearm.
Ordering information: Available in a longer 15" length, see AlphaTec 58-335.