Human Thrombin (from Plasma)
Supplier: MilliporeSigma
605195-1000U
605195-100U
605195-5000U
CA80054-692EA
1088.27
CAD
CA80054-692
CA80054-694
CA80054-696
Human Thrombin (from Plasma)
Enzymes
Designed for use in thrombin clotting time tests.
Key enzyme that activates factor XIII and converts fibrinogen into fibrin. Soluble in aqueous buffers or water. Lyophilized from 200 mm NaCl, 50 mm sodium citrate, 0.1% PEG, pH 6.5.